Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317188.1 | NP_001304117.1 | 5 | 993 | ||
NM_199051.2 | NP_950252.1 | 6 | 1388 | |||
XM_011509472.2 | XP_011507774.1 | 7 | 1390 | |||
XM_011509475.2 | XP_011507777.1 | 6 | 1128 | |||
XM_011509476.2 | XP_011507778.1 | 5 | 1206 | |||
XM_017001125.1 | XP_016856614.1 | 8 | 1499 | |||
XM_017001126.1 | XP_016856615.1 | 7 | 1364 | |||
XM_017001127.1 | XP_016856616.1 | 7 | 1528 | |||
XM_017001128.1 | XP_016856617.1 | 7 | 2616 | |||
XM_017001129.1 | XP_016856618.1 | 5 | 1197 | |||
XM_017001130.1 | XP_016856619.1 | 6 | 1173 | |||
XM_017001131.1 | XP_016856620.1 | 7 | 1478 | |||
XM_017001132.1 | XP_016856621.1 | 6 | 1408 | |||
XM_017001133.1 | XP_016856622.1 | 6 | 1408 | |||
GenBank mRNA | AB111893.1 | - | 6 | 1135 | ||
AK123024.1 | - | 6 | 1078 | |||
AK123637.1 | - | 6 | 1133 | |||
AK294365.1 | - | 5 | 795 | |||
BC105052.1 | - | 5 | 959 | 8144055 | ||
BC105054.1 | - | 5 | 959 | 8144057 |