Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320178.1 | NP_001307107.1 | 6 | 832 | ||
NM_001320179.1 | NP_001307108.1 | 6 | 1126 | |||
NM_198494.2 | NP_940896.2 | 6 | 829 | |||
XM_005270809.4 | XP_005270866.1 | 6 | 1559 | |||
XM_006710605.3 | XP_006710668.1 | 6 | 1539 | |||
XM_006710606.3 | XP_006710669.1 | 6 | 1556 | |||
GenBank mRNA | AK122618.1 | - | 6 | 829 | ||
BC041873.1 | - | 6 | 1542 | 5271835 | ||
BC144174.1 | - | 5 | 578 | |||
CV374692.1 | - | 4 | 405 | |||
CX871163.1 | - | 3 | 215 |