Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042784.1 | NP_001036249.1 | 7, 8 | 1072 | ||
XM_005262974.4 | XP_005263031.1 | 8, 9 | 1199 | |||
XM_006714204.3 | XP_006714267.1 | 8, 9 | 1199 | |||
XM_011531908.2 | XP_011530210.1 | 9, 10 | 1303 | |||
XM_011531909.2 | XP_011530211.1 | 9, 10 | 1319 | |||
XM_011531910.2 | XP_011530212.1 | 8, 9 | 1780 | |||
XM_011531911.1 | XP_011530213.1 | 8, 9 | 1163 | |||
XM_011531912.2 | XP_011530214.1 | 8, 9 | 1059 | |||
XM_011531913.1 | XP_011530215.1 | 7, 8 | 1087 | |||
XM_011531914.2 | XP_011530216.1 | 7, 8 | 1156 | |||
XM_011531915.2 | XP_011530217.1 | 8, 9 | 1200 | |||
XM_011531916.2 | XP_011530218.1 | 7, 8 | 981 | |||
XM_011531917.1 | XP_011530219.1 | 8, 9 | 1024 | |||
XM_017008160.1 | XP_016863649.1 | 9, 10 | 1813 | |||
XM_017008161.1 | XP_016863650.1 | 3, 4 | 264 | |||
GenBank mRNA | AK098636.1 | - | 6, 7 | 879 | ||
BC086869.1 | - | 3, 4 | 442 |