Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004310.2 | NP_001004310.2 | 3 | 172 | ||
NM_001284217.1 | NP_001271146.1 | 4 | 238 | |||
XM_005245128.3 | XP_005245185.1 | 4 | 230 | |||
XM_005245129.3 | XP_005245186.1 | 3 | 197 | |||
XM_005245131.3 | XP_005245188.1 | 4 | 248 | |||
XM_006711292.3 | XP_006711355.1 | 4 | 248 | |||
XM_011509480.1 | XP_011507782.1 | 4 | 237 | |||
XM_011509481.2 | XP_011507783.1 | 4 | 248 | |||
GenBank mRNA | AK131201.1 | - | 4 | 237 | ||
AK301270.1 | - | 3 | 204 | |||
AY212514.1 | - | 3 | 138 | |||
AY513661.1 | - | 3 | 130 | |||
AY654627.1 | - | 3 | 131 | |||
AY654628.1 | - | 3 | 131 | |||
DA959587.1 | - | 3 | 172 |