Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198560.2 | NP_940962.1 | 2 | 646 | ||
XM_017006359.1 | XP_016861848.1 | 1 | 1184 | |||
XM_017006360.1 | XP_016861849.1 | 1 | 1222 | |||
GenBank mRNA | AY278320.1 | - | 2 | 646 | ||
BC113964.1 | - | 2 | 382 |