Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199350.3 | NP_955382.3 | 3 | 880 | ||
XM_011518655.1 | XP_011516957.1 | 5 | 1435 | |||
XM_011518656.1 | XP_011516958.1 | 4 | 945 | |||
XM_011518657.2 | XP_011516959.1 | 4 | 676 | |||
XM_011518658.2 | XP_011516960.1 | 4 | 676 | |||
XM_011518659.1 | XP_011516961.1 | 4 | 676 | |||
XM_011518660.1 | XP_011516962.1 | 4 | 676 | |||
XM_011518663.1 | XP_011516965.1 | 4 | 676 | |||
XM_017014708.1 | XP_016870197.1 | 4 | 676 | |||
GenBank mRNA | AK093122.1 | - | 3 | 880 | ||
JF442034.1 | - | 3 | 351 |