Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201550.3 | NP_963844.2 | 1 | 549 | ||
GenBank mRNA | AK123908.1 | - | 1 | 145 | ||
BC090881.1 | - | 1 | 549 | 30530394 | ||
BC106892.1 | - | 1 | 142 | |||
BC106893.1 | - | 1 | 145 | 40030470 |