Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198582.3 | NP_940984.3 | 2 | 370 | ||
GenBank mRNA | AK125364.1 | - | 1 | 242 | ||
BC103703.1 | - | 1 | 240 | |||
BC104749.1 | - | 1 | 240 | |||
BC105696.1 | - | 1 | 240 | |||
BX280405.1 | - | 2 | 370 |