Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RESP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007089.3 | NP_001007090.3 | 5, 6 | 556 | ||
XM_011511165.2 | XP_011509467.1 | 3, 4 | 419 | |||
XM_011511167.2 | XP_011509469.1 | 4, 5 | 870 | |||
XM_017004079.1 | XP_016859568.1 | 4, 5 | 713 | |||
GenBank mRNA | AF437883.3 | - | 5, 6 | 556 | ||
DQ227570.1 | - | 3, 4 | 627 |