Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201628.2 | NP_963922.2 | 11 | 1953 | ||
XM_005245795.4 | XP_005245852.1 | 13 | 2208 | |||
XM_011541074.2 | XP_011539376.1 | 13 | 2215 | |||
XM_011541075.2 | XP_011539377.1 | 11 | 1871 | |||
XM_011541076.2 | XP_011539378.1 | 11 | 2016 | |||
XM_011541077.2 | XP_011539379.1 | 11 | 2995 | |||
XM_011541081.2 | XP_011539383.1 | 5 | 695 | |||
GenBank mRNA | AK293806.1 | - | 6 | 759 | ||
BC101637.1 | - | 1 | 34 | |||
BC113621.1 | - | 1 | 34 | |||
BC143641.1 | - | 1 | 34 | |||
DN831635.1 | - | 2 | 235 |