Search Thermo Fisher Scientific
Gene Symbol: YY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206923.3 | NP_996806.2 | 1 | 541 | ||
GenBank mRNA | AK091850.1 | - | 1 | 522 | ||
AY567472.1 | - | 1 | 43 | |||
BC137215.1 | - | 1 | 97 | 9021212 | ||
BC137217.1 | - | 1 | 97 | 9021214 | ||
DA961884.1 | - | 1 | 67 | |||
DB029513.1 | - | 1 | 274 |