Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTENP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023917.1 | 1 | 1389 | |||
GenBank mRNA | AF017999.1 | - | 1 | 620 | ||
AF019083.1 | - | 1 | 620 | |||
BC038293.2 | - | 1 | 873 |
Gene Symbol: PTEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000314.6 | NP_000305.3 | 6 | 1651 | ||
NM_001304717.2 | NP_001291646.2 | 6 | 1651 | |||
NM_001304718.1 | NP_001291647.1 | 6 | 1765 | |||
GenBank mRNA | AK313581.1 | - | 6 | 812 | ||
BC005821.2 | - | 6 | 1456 | 3937787 | ||
CR450306.1 | - | 6 | 620 | |||
JF268690.1 | - | 6 | 783 | |||
KJ534922.1 | - | 6 | 620 | |||
U92436.1 | - | 6 | 1654 | |||
U93051.1 | - | 6 | 620 | |||
U96180.1 | - | 6 | 1424 |