Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK32C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318878.1 | NP_001305807.1 | 11 | 1462 | ||
NM_001318879.1 | NP_001305808.1 | 11 | 1323 | |||
NM_173575.3 | NP_775846.2 | 11 | 1424 | |||
NR_134911.1 | 12 | 1171 | ||||
XM_011539688.1 | XP_011537990.1 | 11 | 1495 | |||
XM_011539690.2 | XP_011537992.1 | 11 | 1410 | |||
XM_011539693.2 | XP_011537995.1 | 11 | 1428 | |||
XM_011539694.1 | XP_011537996.1 | 11 | 1162 | |||
XM_011539695.1 | XP_011537997.1 | 11 | 1363 | |||
XM_011539696.1 | XP_011537998.1 | 11 | 1170 | |||
GenBank mRNA | AK057849.1 | - | 12 | 1171 | ||
AK098366.1 | - | 9 | 1451 | |||
AK302102.1 | - | 11 | 1409 | |||
AY098866.1 | - | 11 | 1424 | |||
BC045760.1 | - | 11 | 1313 | 4831007 |