Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003091.3 | NP_003082.1 | 6 | 749 | ||
NM_198216.1 | NP_937859.1 | 6 | 749 | |||
GenBank mRNA | AK301034.1 | - | 7 | 805 | ||
AK301206.1 | - | 6 | 683 | |||
AK312620.1 | - | 6 | 684 | |||
AY742712.1 | - | 5 | 586 | |||
BC080516.1 | - | 6 | 667 | 2822685 | ||
CR456969.1 | - | 5 | 586 | |||
J04564.1 | - | 6 | 709 | |||
M34081.1 | - | 5 | 565 | |||
M34082.1 | - | 5 | 565 | |||
X15893.1 | - | 5 | 586 | |||
X17567.1 | - | 6 | 640 | |||
X17568.1 | - | 6 | 625 |