Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170.2 | NP_001161.1 | 4 | 506 | ||
NM_001318156.1 | NP_001305085.1 | 3 | 388 | |||
NM_001318157.1 | NP_001305086.1 | 4 | 824 | |||
NM_001318158.1 | NP_001305087.1 | 4 | 506 | |||
XM_005251453.3 | XP_005251510.1 | 5 | 606 | |||
XM_006716765.3 | XP_006716828.1 | 3 | 267 | |||
XM_011517866.2 | XP_011516168.1 | 4 | 416 | |||
XM_011517867.2 | XP_011516169.1 | 2 | 46 | |||
XM_017014700.1 | XP_016870189.1 | 4 | 707 | |||
XM_017014706.1 | XP_016870195.1 | 5 | 537 | |||
GenBank mRNA | AB006190.1 | - | 4 | 343 | ||
AK297860.1 | - | 4 | 411 | |||
AK297885.1 | - | 4 | 640 | |||
BC062701.1 | - | 3 | 215 | 30324263 |