Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197216.2 | NP_001184145.1 | 8 | 1117 | ||
NM_001671.4 | NP_001662.1 | 9 | 1234 | |||
XM_011523861.2 | XP_011522163.1 | 9 | 1141 | |||
GenBank mRNA | AB070933.1 | - | 1013 | |||
AK308375.1 | - | 9 | 1038 | |||
AK308677.1 | - | 6 | 992 | |||
BC032130.2 | - | 9 | 911 | 5087444 | ||
BM818592.1 | - | 1 | 34 | |||
BT019873.1 | - | 8 | 834 | |||
CR542052.1 | - | 8 | 834 | |||
FJ410292.1 | - | 7 | 717 | |||
M10058.1 | - | 9 | 1006 |