Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001672.2 | NP_001663.2 | 2, 3 | 232 | ||
XM_005260412.3 | XP_005260469.1 | 3, 4 | 2288 | |||
XM_011528820.2 | XP_011527122.1 | 4, 5 | 411 | |||
XM_011528821.1 | XP_011527123.1 | 4, 5 | 369 | |||
GenBank mRNA | BC104238.1 | - | 2, 3 | 224 | 40033052 | |
BC104239.2 | - | 2, 3 | 224 | 40033056 |