Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007100.3 | NP_009031.1 | 3 | 206 | ||
NR_033743.1 | 2 | 151 | ||||
GenBank mRNA | AB028624.1 | - | 3 | 158 | ||
AK000656.1 | - | 3 | 121 | |||
BC003679.1 | - | 3 | 142 | 4052832 | ||
BC017215.2 | - | 2 | 339 | |||
BC105610.1 | - | 3 | 148 | 6739801 | ||
BF215596.1 | - | 3 | 205 | |||
BG940181.1 | - | 3 | 134 | |||
BG943252.1 | - | 218 | ||||
BT006922.1 | - | 3 | 115 | |||
D50371.1 | - | 3 | 178 | |||
DA119852.1 | - | 2 | 143 |