Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001716.4 | NP_001707.1 | 2 | 347 | ||
NM_032966.2 | NP_116743.1 | 1 | 324 | |||
GenBank mRNA | AK291512.1 | - | 2 | 347 | ||
BC110352.1 | - | 2 | 261 | 5219901 | ||
DA815880.1 | - | 2 | 281 | |||
X68149.1 | - | 2 | 255 | |||
X68829.1 | - | 1 | 324 |