Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000759.3 | NP_000750.1 | 4 | 525 | ||
NM_001178147.1 | NP_001171618.1 | 3 | 408 | |||
NM_172219.2 | NP_757373.1 | 4 | 516 | |||
NM_172220.2 | NP_757374.2 | 3 | 417 | |||
NR_033662.1 | 3 | 692 | ||||
GenBank mRNA | AK290247.1 | - | 3 | 676 | ||
AK298116.1 | - | 2 | 568 | |||
AK313509.1 | - | 4 | 441 | |||
BC033245.1 | - | 3 | 586 | 5434958 | ||
CB127794.1 | - | 3 | 408 | |||
CD013926.1 | - | 3 | 311 | |||
CR541891.1 | - | 4 | 402 | |||
HM132049.1 | - | 2 | 204 | |||
KP271008.1 | - | 3 | 312 | |||
M17706.1 | - | 4 | 444 | |||
X03438.1 | - | 4 | 442 | |||
X03655.1 | - | 4 | 432 |