Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ECI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001919.3 | NP_001910.2 | 5 | 554 | ||
GenBank mRNA | AK291127.1 | - | 5 | 554 | ||
BC000762.2 | - | 5 | 519 | 2821822 | ||
BC009631.1 | - | 6 | 496 | 3877066 | ||
BC019316.1 | - | 5 | 519 | 2821822 | ||
BQ773923.1 | - | 3 | 513 | |||
L24774.1 | - | 4 | 411 | |||
Z25820.1 | - | 5 | 527 |