Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004942.3 | NP_004933.1 | 2 | 250 | ||
GenBank mRNA | BC069285.1 | - | 2 | 138 | 7262691 | |
BC093983.1 | - | 2 | 125 | 7939828 | ||
BC093985.1 | - | 2 | 129 | 7939830 | ||
BM855542.1 | - | 2 | 166 | |||
HY264007.1 | - | 1 | 226 | |||
Z71389.1 | - | 2 | 125 |
Gene Symbol: DEFB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205266.1 | NP_001192195.1 | 2 | 107 | ||
GenBank mRNA | BE714502.1 | - | 2 | 95 | ||
BE714509.1 | - | 1 | 206 |