Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080449.2 | NP_001073918.2 | 16 | 2558 | ||
NR_102264.1 | 17 | 2423 | ||||
XM_006717680.3 | XP_006717743.1 | 17 | 2938 | |||
XM_011539417.1 | XP_011537719.1 | 10 | 1482 | |||
XM_017015799.1 | XP_016871288.1 | 13 | 1947 | |||
GenBank mRNA | AK307939.1 | - | 5 | 509 | ||
AK308358.1 | - | 14 | 2200 | |||
BC028188.1 | - | 3 | 424 | |||
BC041115.2 | - | 4 | 576 | |||
BC053574.1 | - | 6 | 752 | |||
BC073945.1 | - | 2 | 165 | |||
BC111740.1 | - | 17 | 2423 | 40038149 | ||
D42046.1 | - | 16 | 2500 |