Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004467.3 | NP_004458.3 | 8 | 982 | ||
NM_147203.2 | NP_671736.2 | 8 | 1070 | |||
NM_201552.1 | NP_963846.1 | 9 | 1109 | |||
NM_201553.1 | NP_963847.1 | 9 | 1197 | |||
XM_017013232.1 | XP_016868721.1 | 9 | 1959 | |||
XM_017013233.1 | XP_016868722.1 | 7 | 1034 | |||
GenBank mRNA | AF042354.1 | - | 8 | 1018 | ||
AF168954.1 | - | 8 | 1005 | |||
AK225616.1 | - | 8 | 968 | |||
AK225619.1 | - | 8 | 957 | |||
AK289448.1 | - | 8 | 968 | |||
BC007047.1 | - | 8 | 978 | 4071810 | ||
BT006635.1 | - | 7 | 873 | |||
D14446.1 | - | 8 | 951 | |||
D87342.1 | - | 7 | 878 |