Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039492.2 | NP_001034581.1 | 6 | 860 | ||
NM_001318894.1 | NP_001305823.1 | 5 | 1106 | |||
NM_001318895.1 | NP_001305824.1 | 6 | 1094 | |||
NM_001318896.1 | NP_001305825.1 | 5 | 809 | |||
NM_001318897.1 | NP_001305826.1 | 4 | 860 | |||
NM_001318898.1 | NP_001305827.1 | 5 | 919 | |||
NM_001318899.1 | NP_001305828.1 | 7 | 982 | |||
NM_001450.3 | NP_001441.4 | 5 | 1043 | |||
NM_201555.1 | NP_963849.1 | 7 | 1215 | |||
NM_201557.3 | NP_963851.2 | 5 | 1035 | |||
XM_011510798.2 | XP_011509100.1 | 6 | 842 | |||
GenBank mRNA | AB158503.1 | - | 4 | 582 | ||
AK313841.1 | - | 6 | 892 | |||
AL521653.3 | - | 5 | 736 | |||
AY634685.1 | - | 4 | 636 | |||
BC012742.2 | - | 7 | 1215 | 3840517 | ||
BC014397.1 | - | 7 | 1215 | 3546686 | ||
BC036085.1 | - | 5 | 690 | |||
BC093049.1 | - | 5 | 801 | 30374919 | ||
BI092736.1 | - | 2 | 153 | |||
BT006960.1 | - | 4 | 636 | |||
CF552311.1 | - | 5 | 766 | |||
CR457437.1 | - | 4 | 636 | |||
CR936682.1 | - | 5 | 1106 | |||
CR936799.1 | - | 5 | 6087 | |||
DQ266432.1 | - | 6 | 1094 | |||
DQ307067.1 | - | 5 | 973 | |||
JQ313902.1 | - | 4 | 606 | |||
L42176.1 | - | 6 | 774 | |||
U29332.1 | - | 5 | 757 | |||
U60117.1 | - | 2 | 259 |