Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319077.1 | NP_001306006.1 | 6 | 1567 | ||
NM_001319078.1 | NP_001306007.1 | 5 | 1488 | |||
NM_001319079.1 | NP_001306008.1 | 6 | 1600 | |||
NM_001319080.1 | NP_001306009.1 | 6 | 1599 | |||
NM_001319081.1 | NP_001306010.1 | 5 | 1486 | |||
NM_001319082.1 | NP_001306011.1 | 6 | 1622 | |||
NM_002131.3 | NP_002122.1 | 6 | 1569 | |||
NM_145899.2 | NP_665906.1 | 6 | 1602 | |||
NM_145901.2 | NP_665908.1 | 5 | 1769 | |||
NM_145902.2 | NP_665909.1 | 5 | 1736 | |||
NM_145903.2 | NP_665910.1 | 5 | 1455 | |||
NM_145905.2 | NP_665912.1 | 5 | 1458 | |||
GenBank mRNA | AK096863.1 | - | 7 | 1509 | ||
AK301434.1 | - | 5 | 1462 | |||
BC004924.2 | - | 6 | 1449 | 3605116 | ||
BC008832.2 | - | 6 | 1482 | 4308914 | ||
BC015789.1 | - | 5 | 1458 | 4866014 | ||
BC063434.1 | - | 5 | 1374 | 5399570 | ||
BC067083.1 | - | 6 | 1457 | 4110270 | ||
BC071863.1 | - | 4 | 1062 | 4651720 | ||
BC071864.1 | - | 6 | 1477 | 6083428 | ||
M23614.1 | - | 5 | 1486 | |||
M23615.1 | - | 5 | 1453 | |||
M23616.1 | - | 6 | 1488 | |||
M23617.1 | - | 5 | 1381 | |||
M23618.1 | - | 4 | 1288 | |||
M23619.1 | - | 6 | 1591 | |||
X14957.1 | - | 6 | 1512 | |||
X14958.1 | - | 6 | 1479 |