Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HMGA1 SIRNA Assays ›
Gene Symbol:
HMGA1
Gene Name:
high mobility group AT-hook 1
Gene Aliases:
HMG-R, HMGA1A, HMGIY
Chromosome Location:
Chr.6: 34236800 - 34246231 on Build GRCh38
UniGene ID:
Hs.518805
Species:
Human
siRNA ID
HSS142461
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HMGA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319077.1
NP_001306006.1
6
577
NM_001319078.1
NP_001306007.1
5
498
NM_001319079.1
NP_001306008.1
6
610
NM_001319080.1
NP_001306009.1
6
609
NM_001319081.1
NP_001306010.1
5
496
NM_001319082.1
NP_001306011.1
6
632
NM_002131.3
NP_002122.1
6
579
NM_145899.2
NP_665906.1
6
612
NM_145901.2
NP_665908.1
5
779
NM_145902.2
NP_665909.1
5
746
NM_145903.2
NP_665910.1
5
465
NM_145905.2
NP_665912.1
5
468
GenBank mRNA
AF176039.1
-
4
284
AK096863.1
-
7
519
AK301434.1
-
5
472
AK308245.1
-
5
753
BC004924.2
-
6
459
3605116
BC008832.2
-
6
492
4308914
BC015789.1
-
5
468
4866014
BC063434.1
-
5
384
5399570
BC067083.1
-
6
467
4110270
BC071864.1
-
6
487
6083428
BE269847.1
-
6
493
BT006774.1
-
4
257
CX785762.1
-
5
400
DA467015.1
-
6
538
DA574958.1
-
5
447
DA936387.1
-
6
535
DB029421.1
-
6
560
M23614.1
-
5
497
M23615.1
-
5
464
M23616.1
-
6
499
M23617.1
-
5
392
M23618.1
-
4
299
M23619.1
-
6
602
X14957.1
-
6
521
X14958.1
-
6
488
Z36875.1
-
11
Back To Top
More Information
Panther Classification:
Molecular Function -
endodeoxyribonuclease
Gene Ontology Categories:
Function(s)
Process(es)
DNA unwinding involved in DNA replication
nucleosome disassembly
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
negative regulation of cell proliferation
response to virus
negative regulation of chromatin silencing
senescence-associated heterochromatin focus assembly
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nuclear transport
establishment of integrated proviral latency
oncogene-induced cell senescence
positive regulation of cellular senescence
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
AT DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
enzyme binding
ligand-dependent nuclear receptor transcription coactivator activity
retinoic acid receptor binding
peroxisome proliferator activated receptor binding
retinoid X receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-d4wl8:80/10.239.162.132:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0