Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002133.2 | NP_002124.1 | 3 | 724 | ||
GenBank mRNA | AF051791.1 | - | 2 | 597 | ||
AK313120.1 | - | 3 | 694 | |||
BC001491.2 | - | 3 | 683 | 3504480 | ||
BT019785.1 | - | 2 | 617 | |||
CR456505.1 | - | 3 | 732 | |||
CR541945.1 | - | 3 | 617 | |||
CR541968.1 | - | 3 | 617 | |||
JF323038.1 | - | 3 | 621 | |||
X06985.1 | - | 3 | 697 |