Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HPS1 SIRNA Assays ›
Gene Symbol:
HPS1
Gene Name:
HPS1, biogenesis of lysosomal organelles complex 3 subunit 1
Gene Aliases:
BLOC3S1, HPS
Chromosome Location:
Chr.10: 98415068 - 98446963 on Build GRCh38
UniGene ID:
Hs.404568
Species:
Human
siRNA ID
HSS142541
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HPS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000195.4
NP_000186.2
3
323
NM_001311345.1
NP_001298274.1
3
332
NM_001322476.1
NP_001309405.1
3
252
NM_001322477.1
NP_001309406.1
3
332
NM_001322478.1
NP_001309407.1
3
252
NM_001322479.1
NP_001309408.1
3
332
NM_001322480.1
NP_001309409.1
3
252
NM_001322481.1
NP_001309410.1
3
332
NM_001322482.1
NP_001309411.1
3
332
NM_001322483.1
NP_001309412.1
3
252
NM_001322484.1
NP_001309413.1
3
332
NM_001322485.1
NP_001309414.1
3
323
NM_001322487.1
NP_001309416.1
3
332
NM_001322489.1
NP_001309418.1
3
332
NM_001322490.1
NP_001309419.1
3
332
NM_001322491.1
NP_001309420.1
3
252
NM_001322492.1
NP_001309421.1
3
252
NM_182639.3
NP_872577.1
3
332
XM_005269757.4
XP_005269814.1
3
265
XM_017016170.1
XP_016871659.1
3
239
XM_017016171.1
XP_016871660.1
3
328
XM_017016172.1
XP_016871661.1
3
300
XM_017016173.1
XP_016871662.1
3
330
GenBank mRNA
AK295821.1
-
3
288
AL702912.1
-
3
317
BC000175.2
-
3
282
BI518436.1
-
3
242
DA385131.1
-
3
330
DA479757.1
-
3
252
KU178114.1
-
1
61
KU178115.1
-
1
61
KU178116.1
-
1
61
KU178117.1
-
1
61
KU178118.1
-
1
61
KU178119.1
-
1
61
KU178120.1
-
1
61
KU178121.1
-
1
61
KU178122.1
-
1
61
U65676.1
-
3
267
U96721.1
-
3
295
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lysosome organization
visual perception
response to stimulus
protein binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-vrvhj:80/10.239.161.228:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0