Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303233.1 | NP_001290162.1 | 3 | 423 | ||
NM_001303234.1 | NP_001290163.1 | 4 | 549 | |||
NM_001303235.1 | NP_001290164.1 | 2 | 2046 | |||
NM_001303236.1 | NP_001290165.1 | 3 | 2670 | |||
NM_001303237.1 | NP_001290166.1 | 3 | 633 | |||
NM_002201.5 | NP_002192.2 | 3 | 693 | |||
NR_130134.1 | 2 | 171 | ||||
XM_005254899.2 | XP_005254956.1 | 3 | 1956 | |||
XM_006720488.3 | XP_006720551.1 | 5 | 734 | |||
XM_011521521.2 | XP_011519823.1 | 2 | 1116 | |||
XM_017022147.1 | XP_016877636.1 | 3 | 3757 | |||
XM_017022148.1 | XP_016877637.1 | 4 | 512 | |||
GenBank mRNA | AB209639.1 | - | 1 | 5305 | ||
AK122793.1 | - | 2 | 2010 | |||
AK307769.1 | - | 3 | 491 | |||
AV737297.1 | - | 2 | 193 | |||
BC007922.2 | - | 3 | 628 | 4309120 | ||
BC016341.1 | - | 3 | 2371 | |||
BI838264.1 | - | 4 | 502 | |||
BM691295.1 | - | 3 | 431 | |||
BT006952.1 | - | 2 | 335 | |||
BX339191.2 | - | 3 | 631 | |||
CN265233.1 | - | 3 | 423 | |||
CR456942.1 | - | 2 | 335 | |||
U88964.1 | - | 2 | 372 | |||
X89773.2 | - | 2 | 350 |