- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: KRT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000526.4 | NP_000517.2 | 8 | 1561 | ||
GenBank mRNA | BC002690.2 | - | 8 | 1546 | 3610489 | |
BC019097.2 | - | 8 | 1552 | 5013623 | ||
BC042437.1 | - | 8 | 1516 | 4776763 | ||
BC094830.1 | - | 8 | 1552 | 5443153 |