Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166386.3 | NP_001159858.1 | 3 | 1693 | ||
NM_001166387.3 | NP_001159859.1 | 3 | 1601 | |||
NM_005367.6 | NP_005358.2 | 2 | 1495 | |||
GenBank mRNA | BC003408.2 | - | 2 | 1483 | 3450217 |