Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012333.2 | NP_001012333.1 | 4, 5 | 587 | ||
NM_001012334.2 | NP_001012334.1 | 4, 5 | 633 | |||
NM_001270550.1 | NP_001257479.1 | 4, 5 | 812 | |||
NM_001270551.1 | NP_001257480.1 | 3, 4 | 758 | |||
NM_001270552.1 | NP_001257481.1 | 3, 4 | 348 | |||
NM_002391.4 | NP_002382.1 | 4, 5 | 516 | |||
NR_073039.1 | 3, 4 | 439 | ||||
XM_011520116.2 | XP_011518418.1 | 4, 5 | 799 | |||
XM_017017764.1 | XP_016873253.1 | 4, 5 | 655 | |||
GenBank mRNA | AK130889.1 | - | 4, 5 | 633 | ||
BC011704.2 | - | 4, 5 | 447 | 3609004 | ||
BF684205.1 | - | 3, 4 | 377 | |||
BX346163.2 | - | 3, 4 | 716 | |||
CN266354.1 | - | 3, 4 | 401 | |||
CN266369.1 | - | 4, 5 | 508 | |||
DQ323888.1 | - | 3, 4 | 334 | |||
DQ323889.1 | - | 3, 4 | 257 | |||
DQ323890.1 | - | 4, 5 | 259 | |||
M69148.1 | - | 3, 4 | 411 | |||
X55110.1 | - | 4, 5 | 436 |