Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MITF SIRNA Assays ›
Gene Symbol:
MITF
Gene Name:
melanogenesis associated transcription factor
Gene Aliases:
CMM8, MI, WS2, WS2A, bHLHe32
Chromosome Location:
Chr.3: 69739435 - 69968337 on Build GRCh38
UniGene ID:
Hs.166017
Species:
Human
siRNA ID
HSS142940
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MITF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000248.3
NP_000239.1
9
1259
NM_001184967.1
NP_001171896.1
10
1472
NM_006722.2
NP_006713.1
10
1467
NM_198158.2
NP_937801.1
9
1241
NM_198159.2
NP_937802.1
10
1602
NM_198177.2
NP_937820.1
10
1458
NM_198178.2
NP_937821.2
9
1073
XM_005264754.1
XP_005264811.1
10
1592
XM_005264755.3
XP_005264812.1
10
1618
XM_006713164.2
XP_006713227.1
11
1612
XM_011533722.2
XP_011532024.1
10
1597
XM_011533723.2
XP_011532025.1
10
1617
XM_011533725.1
XP_011532027.1
10
1452
XM_011533726.1
XP_011532028.1
10
1434
XM_017006444.1
XP_016861933.1
10
9807
XM_017006445.1
XP_016861934.1
10
1579
XM_017006446.1
XP_016861935.1
10
1459
XM_017006447.1
XP_016861936.1
11
1710
XM_017006448.1
XP_016861937.1
11
1594
GenBank mRNA
AB006909.1
-
10
1555
AK291318.1
-
10
1574
AK296129.1
-
10
1472
AK297858.1
-
10
1404
AL110195.1
-
10
1558
BC026961.2
-
9
1171
3924101
BC065243.1
-
9
1216
6066096
GU355676.1
-
9
950
Z29678.1
-
9
1256
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein complex assembly
multicellular organism development
positive regulation of gene expression
protein sumoylation
melanocyte differentiation
regulation of apoptotic process
regulation of osteoclast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
mast cell migration
positive regulation of DNA-templated transcription, initiation
regulation of RNA biosynthetic process
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
protein binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0