Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013436.2 | NP_001013454.1 | 3 | 866 | ||
NM_001130517.2 | NP_001123989.1 | 4 | 1042 | |||
NM_021126.5 | NP_066949.2 | 3 | 856 | |||
XM_005261610.3 | XP_005261667.1 | 4 | 987 | |||
XM_011530195.1 | XP_011528497.1 | 4 | 906 | |||
GenBank mRNA | AB208824.1 | - | 3 | 5390 | ||
AK055733.1 | - | 3 | 1624 | |||
BC003508.1 | - | 2 | 739 | 3611253 | ||
BC009450.1 | - | 2 | 739 | |||
BC016737.1 | - | 3 | 829 | 4105509 | ||
BC018717.1 | - | 2 | 736 | 4131927 | ||
BT019636.1 | - | 2 | 709 | |||
CR456523.1 | - | 2 | 728 | |||
CR541712.1 | - | 2 | 709 | |||
CX866643.1 | - | 3 | 749 | |||
X59434.1 | - | 2 | 740 |