Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MUC1 SIRNA Assays ›
Gene Symbol:
MUC1
Gene Name:
mucin 1, cell surface associated
Gene Aliases:
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
Chromosome Location:
Chr.1: 155185824 - 155192915 on Build GRCh38
UniGene ID:
Hs.89603
Species:
Human
siRNA ID
HSS142992
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MUC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001018016.2
NP_001018016.1
8
812
NM_001018017.2
NP_001018017.1
8
785
NM_001044390.2
NP_001037855.1
7
629
NM_001044391.2
NP_001037856.1
5
470
NM_001044392.2
NP_001037857.1
5
497
NM_001044393.2
NP_001037858.1
7
597
NM_001204285.1
NP_001191214.1
8
1445
NM_001204286.1
NP_001191215.1
8
1472
NM_001204287.1
NP_001191216.1
8
866
NM_001204288.1
NP_001191217.1
8
780
NM_001204289.1
NP_001191218.1
7
734
NM_001204290.1
NP_001191219.1
7
671
NM_001204291.1
NP_001191220.1
7
743
NM_001204292.1
NP_001191221.1
7
737
NM_001204293.1
NP_001191222.1
7
717
NM_001204294.1
NP_001191223.1
7
710
NM_001204295.1
NP_001191224.1
6
587
NM_001204296.1
NP_001191225.1
7
656
NM_001204297.1
NP_001191226.1
5
551
NM_002456.5
NP_002447.4
8
839
GenBank mRNA
AF125525.1
-
8
704
AF348143.1
-
5
476
AK301575.1
-
9
1697
AY327582.1
-
7
581
AY327583.1
-
5
422
AY327584.1
-
5
449
AY327585.1
-
5
443
AY327586.1
-
5
470
AY327587.1
-
8
790
AY327588.1
-
7
685
AY327589.1
-
5
475
AY327591.1
-
7
580
AY327592.1
-
7
600
AY327596.1
-
9
920
AY327597.1
-
6
459
AY327598.1
-
7
549
AY327599.1
-
6
552
AY327600.1
-
7
568
BC120974.2
-
8
762
40115473
BC120975.2
-
8
789
40115475
BM969296.1
-
1
379
BM971650.1
-
1
379
EF583642.1
-
8
1115
EF583643.1
-
8
943
EF583644.1
-
8
929
EF583645.1
-
8
889
EF583646.1
-
8
857
EF583647.1
-
8
844
EF583648.1
-
8
771
EF583649.1
-
8
758
EF583650.1
-
8
727
EF583651.1
-
8
722
EF583652.1
-
7
712
EF583653.1
-
7
670
EF583654.1
-
7
649
EF583655.1
-
6
597
EF583656.1
-
5
458
EF670706.1
-
8
1139
EF670707.1
-
8
808
EF670708.1
-
7
721
EF670709.1
-
7
715
EF670710.1
-
7
707
EF670711.1
-
7
695
EF670712.1
-
7
688
EF670713.1
-
7
653
EF670714.1
-
7
643
EF670715.1
-
5
637
EF670716.1
-
6
565
EF670717.1
-
6
550
EF670718.1
-
5
529
EF670719.1
-
5
499
EF670720.1
-
4
459
EF670722.1
-
5
433
EF670723.1
-
3
229
FJ226028.1
-
8
1408
FJ226029.1
-
7
1404
FJ226030.1
-
8
1374
FJ226031.1
-
7
1332
FJ226032.1
-
7
1265
FJ226033.1
-
7
1246
FJ226034.1
-
8
1165
FJ226035.1
-
7
1148
FJ226036.1
-
6
1128
FJ226037.1
-
9
1105
FJ226038.1
-
5
1071
FJ226039.1
-
9
1032
FJ226040.1
-
9
952
FJ226041.1
-
9
947
FJ226042.1
-
9
919
FJ226043.1
-
8
830
FJ226044.1
-
7
814
FJ226045.1
-
7
784
FJ226046.1
-
7
779
FJ226047.1
-
8
744
FJ226049.1
-
7
707
FJ226050.1
-
7
680
FJ226051.1
-
7
653
FJ226052.1
-
7
641
FJ226053.1
-
7
634
FJ226054.1
-
6
630
FJ226055.1
-
6
578
J05581.1
-
8
1445
M34088.1
-
6
932
M34089.1
-
6
932
U60259.1
-
7
663
U60260.1
-
8
738
U60261.1
-
8
792
X52229.1
-
9
1550
X80761.1
-
8
713
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
negative regulation of transcription by competitive promoter binding
O-glycan processing
negative regulation of cell adhesion mediated by integrin
positive regulation of transcription from RNA polymerase II promoter in response to stress
regulation of transcription from RNA polymerase II promoter in response to stress
positive regulation of histone H4 acetylation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
p53 binding
transcription cofactor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-b8dk7:80/10.239.160.22:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0