Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021077.3 | NP_066563.2 | 3 | 847 | ||
NM_205858.1 | NP_995580.1 | 3 | 842 | |||
GenBank mRNA | AL110112.1 | - | 3 | 466 | ||
BC007407.2 | - | 3 | 835 | 3030430 | ||
BC007431.2 | - | 3 | 835 | 3030430 | ||
BC008603.1 | - | 3 | 467 | 4185578 | ||
M21551.1 | - | 3 | 465 |