Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OGG1 SIRNA Assays ›
Gene Symbol:
OGG1
Gene Name:
8-oxoguanine DNA glycosylase
Gene Aliases:
HMMH, HOGG1, MUTM, OGH1
Chromosome Location:
Chr.3: 9749944 - 9788246 on Build GRCh38
UniGene ID:
Hs.380271
Species:
Human
siRNA ID
HSS143187
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OGG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002542.5
NP_002533.1
2, 3
733
NM_016819.3
NP_058212.1
2, 3
733
NM_016820.3
NP_058213.1
2, 3
733
NM_016821.2
NP_058214.1
2, 3
733
NM_016826.2
NP_058434.1
2, 3
733
NM_016827.2
NP_058436.1
2, 3
733
NM_016828.2
NP_058437.1
2, 3
733
NM_016829.2
NP_058438.1
2, 3
733
XM_011533760.1
XP_011532062.1
2, 3
728
XM_017006493.1
XP_016861982.1
2, 3
728
XM_017006494.1
XP_016861983.1
2, 3
728
XM_017006495.1
XP_016861984.1
2, 3
728
XM_017006496.1
XP_016861985.1
2, 3
728
XM_017006497.1
XP_016861986.1
2, 3
728
XM_017006498.1
XP_016861987.1
2, 3
728
XM_017006499.1
XP_016861988.1
2, 3
728
GenBank mRNA
AB000410.1
-
2, 3
658
AB019528.1
-
1, 2
106
AB019529.1
-
1, 2
106
AB019530.1
-
1, 2
149
AB019531.1
-
1, 2
149
AF003595.1
-
2, 3
446
AF026691.1
-
2, 3
597
AK289858.1
-
2, 3
686
BC000657.2
-
2, 3
687
3350168
U88527.1
-
2, 3
700
U88620.1
-
2, 3
700
U96710.1
-
2, 3
447
Y11731.2
-
2, 3
698
Y11838.1
-
2, 3
697
Y13277.1
-
2, 3
694
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
acute inflammatory response
base-excision repair
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
response to oxidative stress
aging
response to radiation
response to light stimulus
response to estradiol
regulation of protein import into nucleus, translocation
nucleotide-excision repair, DNA incision
response to drug
negative regulation of apoptotic process
depurination
depyrimidination
response to ethanol
response to folic acid
cellular response to cadmium ion
negative regulation of double-strand break repair via single-strand annealing
damaged DNA binding
endonuclease activity
protein binding
microtubule binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
oxidized purine DNA binding
8-oxo-7,8-dihydroguanine DNA N-glycosylase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-q6sk2:80/10.239.167.246:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline