Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031848.1 | NP_001027018.1 | 3, 4 | 466 | ||
NM_001276490.1 | NP_001263419.1 | 2, 3 | 288 | |||
NM_002640.3 | NP_002631.3 | 3, 4 | 466 | |||
NM_198833.1 | NP_942130.1 | 3, 4 | 483 | |||
XM_006722484.2 | XP_006722547.1 | 3, 4 | 414 | |||
XM_011526025.1 | XP_011524327.1 | 3, 4 | 467 | |||
XM_017025790.1 | XP_016881279.1 | 3, 4 | 467 | |||
XM_017025791.1 | XP_016881280.1 | 3, 4 | 411 | |||
XM_017025792.1 | XP_016881281.1 | 2, 3 | 233 | |||
GenBank mRNA | AK300391.1 | - | 2, 3 | 236 | ||
AK313782.1 | - | 3, 4 | 383 | |||
AL708543.1 | - | 3, 4 | 431 | |||
BC034528.1 | - | 3, 4 | 376 | 4780455 | ||
BQ435307.1 | - | 3, 4 | 489 | |||
BX571754.1 | - | 3, 4 | 374 | |||
DC384755.1 | - | 2, 3 | 222 | |||
DQ656055.2 | - | 3, 4 | 360 | |||
L40377.1 | - | 3, 4 | 397 | |||
U92984.1 | - | 3, 4 | 375 |