Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002641.3 | NP_002632.1 | 6 | 1517 | ||
NM_020473.3 | NP_065206.3 | 5 | 740 | |||
NR_033835.1 | 6 | 1143 | ||||
NR_033836.1 | 6 | 859 | ||||
XM_011545539.2 | XP_011543841.1 | 5 | 2341 | |||
XM_017029581.1 | XP_016885070.1 | 5 | 1464 | |||
GenBank mRNA | AK098047.1 | - | 5 | 953 | ||
AK303538.1 | - | 5 | 732 | |||
AK315063.1 | - | 5 | 1476 | |||
BC038236.1 | - | 5 | 1474 | 5733899 | ||
D11466.1 | - | 6 | 1486 | |||
S74936.1 | - | 4 | 959 |