Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004581.5 | NP_004572.3 | 2 | 531 | ||
NM_182836.2 | NP_878256.1 | 3 | 214 | |||
GenBank mRNA | AK291347.1 | - | 3 | 98 | ||
AK292613.1 | - | 2 | 263 | |||
AK308715.1 | - | 3 | 97 | |||
AL547259.3 | - | 3 | 188 | |||
AU125148.1 | - | 2 | 441 | |||
BC003093.1 | - | 3 | 152 | 3537388 | ||
CB995791.1 | - | 3 | 211 | |||
CR541797.1 | - | 1 | 19 | |||
DA379718.1 | - | 3 | 214 | |||
DB116988.1 | - | 2 | 482 | |||
Y08200.1 | - | 2 | 293 |