Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002974.3 | NP_002965.1 | 6, 7 | 750 | ||
XM_011526138.1 | XP_011524440.1 | 6, 7 | 760 | |||
GenBank mRNA | AF461147.1 | - | 5, 6 | 613 | ||
AK292212.1 | - | 6, 7 | 750 | |||
AY245780.1 | - | 5, 6 | 613 | |||
AY245782.1 | - | 5, 6 | 613 | |||
BC017401.1 | - | 6, 7 | 690 | 4047303 | ||
U19557.1 | - | 6, 7 | 663 | |||
X89015.1 | - | 5, 6 | 613 |
Gene Symbol: SERPINB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF461146.1 | - | 5, 6 | 613 |