Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST3GAL3 SIRNA Assays ›
Gene Symbol:
ST3GAL3
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases:
EIEE15, MRT12, SIAT6, ST3GALII, ST3GalIII, ST3N
Chromosome Location:
Chr.1: 43707497 - 43931165 on Build GRCh38
UniGene ID:
Hs.597915
Species:
Human
siRNA ID
HSS143939
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST3GAL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270459.1
NP_001257388.1
12
1190
NM_001270460.1
NP_001257389.1
11
1187
NM_001270461.1
NP_001257390.1
9
938
NM_001270462.1
NP_001257391.1
8
845
NM_001270463.1
NP_001257392.1
8
796
NM_001270464.1
NP_001257393.1
8
751
NM_001270465.1
NP_001257394.1
8
822
NM_001270466.1
NP_001257395.1
5
496
NM_006279.3
NP_006270.1
12
1280
NM_174963.3
NP_777623.2
13
1487
NM_174964.2
NP_777624.1
12
1325
NM_174965.2
NP_777625.1
7
684
NM_174966.2
NP_777626.1
10
986
NM_174967.2
NP_777627.1
7
639
NM_174968.3
NP_777628.2
13
1442
NM_174969.2
NP_777629.1
11
1232
NM_174970.2
NP_777630.1
6
591
NM_174971.3
NP_777631.2
12
1394
NR_073016.1
12
1312
NR_073017.1
10
1189
NR_073018.1
11
1159
NR_073019.1
9
1128
NR_073020.1
10
1045
NR_073021.1
10
973
NR_073023.1
9
933
XM_006710825.3
XP_006710888.1
13
1512
XM_006710827.3
XP_006710890.1
12
1430
XM_006710828.3
XP_006710891.1
14
1652
XM_011541974.2
XP_011540276.1
14
1640
XM_011541976.2
XP_011540278.1
13
1592
XM_011541977.2
XP_011540279.1
13
1523
XM_011541980.2
XP_011540282.1
10
1743
XM_011541981.2
XP_011540283.1
11
1195
XM_011541983.2
XP_011540285.1
14
1787
XM_011541987.2
XP_011540289.1
14
1651
XM_011541988.2
XP_011540290.1
9
1668
XM_017002109.1
XP_016857598.1
14
1748
XM_017002111.1
XP_016857600.1
14
1741
XM_017002112.1
XP_016857601.1
14
1737
XM_017002113.1
XP_016857602.1
12
1360
XM_017002114.1
XP_016857603.1
14
1753
XM_017002115.1
XP_016857604.1
13
1604
XM_017002116.1
XP_016857605.1
13
1575
XM_017002117.1
XP_016857606.1
11
1150
XM_017002118.1
XP_016857607.1
10
1033
XM_017002119.1
XP_016857608.1
13
1454
XM_017002120.1
XP_016857609.1
13
1452
XM_017002121.1
XP_016857610.1
11
1245
XM_017002122.1
XP_016857611.1
13
1611
GenBank mRNA
AF425851.1
-
11
1134
AF425852.1
-
6
493
AF425853.1
-
12
1296
AF425854.1
-
11
999
AF425855.1
-
10
996
AF425856.1
-
9
795
AF425857.1
-
6
448
AF425858.1
-
12
1251
AF425859.1
-
10
1041
AF425860.1
-
5
400
AF425861.1
-
11
1203
AF425862.1
-
11
1089
AF425863.1
-
7
631
AF425864.1
-
11
1121
AF425865.1
-
10
968
AF425866.1
-
8
634
AF425867.1
-
9
782
AF425868.1
-
9
998
AF425869.1
-
8
937
AY167992.1
-
8
742
AY167993.1
-
8
747
AY167994.1
-
7
605
AY167995.1
-
7
560
AY167996.1
-
9
854
AY167997.1
-
8
654
AY167998.1
-
4
305
BC050380.1
-
12
1264
5769575
L23768.1
-
11
1089
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
oligosaccharide metabolic process
O-glycan processing
keratan sulfate biosynthetic process
protein N-linked glycosylation via asparagine
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
N-acetyllactosaminide alpha-2,3-sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0