Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST3GAL3 SIRNA Assays ›
Gene Symbol:
ST3GAL3
Gene Name:
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Gene Aliases:
EIEE15, MRT12, SIAT6, ST3GALII, ST3GalIII, ST3N
Chromosome Location:
Chr.1: 43707497 - 43931165 on Build GRCh38
UniGene ID:
Hs.597915
Species:
Human
siRNA ID
HSS143940
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST3GAL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270459.1
NP_001257388.1
2
204
NM_001270460.1
NP_001257389.1
2
204
NM_001270461.1
NP_001257390.1
2
204
NM_001270462.1
NP_001257391.1
2
204
NM_001270463.1
NP_001257392.1
2
204
NM_001270464.1
NP_001257393.1
2
204
NM_001270465.1
NP_001257394.1
2
204
NM_001270466.1
NP_001257395.1
2
204
NM_006279.3
NP_006270.1
2
204
NM_174963.3
NP_777623.2
2
204
NM_174964.2
NP_777624.1
2
204
NM_174965.2
NP_777625.1
2
204
NM_174966.2
NP_777626.1
2
204
NM_174967.2
NP_777627.1
2
204
NM_174968.3
NP_777628.2
2
204
NM_174969.2
NP_777629.1
2
204
NM_174970.2
NP_777630.1
2
204
NM_174971.3
NP_777631.2
2
204
NR_073016.1
2
204
NR_073017.1
2
204
NR_073018.1
2
204
NR_073019.1
2
204
NR_073020.1
2
204
NR_073021.1
2
204
NR_073023.1
2
204
XM_006710825.3
XP_006710888.1
2
240
XM_006710827.3
XP_006710890.1
2
206
XM_006710828.3
XP_006710891.1
2
206
XM_011541973.2
XP_011540275.1
2
206
XM_011541974.2
XP_011540276.1
2
206
XM_011541976.2
XP_011540278.1
2
206
XM_011541977.2
XP_011540279.1
2
206
XM_011541981.2
XP_011540283.1
2
206
XM_011541983.2
XP_011540285.1
2
206
XM_011541984.2
XP_011540286.1
2
206
XM_011541985.2
XP_011540287.1
2
206
XM_011541986.2
XP_011540288.1
2
206
XM_011541987.2
XP_011540289.1
2
206
XM_017002109.1
XP_016857598.1
2
269
XM_017002110.1
XP_016857599.1
2
206
XM_017002111.1
XP_016857600.1
1
43
XM_017002112.1
XP_016857601.1
1
43
XM_017002113.1
XP_016857602.1
2
284
XM_017002114.1
XP_016857603.1
1
43
XM_017002115.1
XP_016857604.1
2
206
XM_017002116.1
XP_016857605.1
1
43
XM_017002117.1
XP_016857606.1
2
206
XM_017002118.1
XP_016857607.1
2
206
XM_017002119.1
XP_016857608.1
2
204
XM_017002120.1
XP_016857609.1
2
203
XM_017002121.1
XP_016857610.1
1
44
XM_017002122.1
XP_016857611.1
1
43
GenBank mRNA
AF425851.1
-
1
13
AF425852.1
-
1
13
AF425853.1
-
1
13
AF425854.1
-
1
13
AF425855.1
-
1
13
AF425856.1
-
1
13
AF425857.1
-
1
13
AF425858.1
-
1
13
AF425859.1
-
1
13
AF425860.1
-
1
13
AF425861.1
-
1
13
AF425862.1
-
1
13
AF425863.1
-
1
13
AF425864.1
-
1
13
AF425865.1
-
1
13
AF425866.1
-
1
13
AF425867.1
-
1
13
AF425868.1
-
1
13
AF425869.1
-
1
13
AK301829.1
-
2
190
AK310265.1
-
2
218
AY051143.1
-
2
189
AY167992.1
-
1
13
AY167993.1
-
1
13
AY167994.1
-
1
13
AY167995.1
-
1
13
AY167996.1
-
1
13
AY167997.1
-
1
13
AY167998.1
-
1
13
BC050380.1
-
2
188
5769575
L23768.1
-
1
13
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
oligosaccharide metabolic process
O-glycan processing
keratan sulfate biosynthetic process
protein N-linked glycosylation via asparagine
sialylation
beta-galactoside (CMP) alpha-2,3-sialyltransferase activity
N-acetyllactosaminide alpha-2,3-sialyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qskjf:80/10.239.165.3:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0