Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006931.2 | NP_008862.1 | 4 | 573 | ||
GenBank mRNA | AB209607.1 | - | 4 | 444 | ||
AK298673.1 | - | 3 | 270 | |||
AK304509.1 | - | 4 | 276 | |||
AK312386.1 | - | 4 | 464 | |||
BC039196.1 | - | 4 | 448 | 4396508 | ||
CR457413.1 | - | 3 | 311 | |||
M20681.1 | - | 4 | 553 |