Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258423.1 | NP_001245352.1 | 3 | 520 | ||
NM_004595.4 | NP_004586.2 | 5 | 679 | |||
XM_005274582.2 | XP_005274639.1 | 5 | 745 | |||
XM_011545568.2 | XP_011543870.1 | 5 | 539 | |||
XM_017029753.1 | XP_016885242.1 | 5 | 519 | |||
XM_017029754.1 | XP_016885243.1 | 5 | 486 | |||
XM_017029755.1 | XP_016885244.1 | 5 | 577 | |||
GenBank mRNA | AD001528.1 | - | 5 | 528 | ||
AK293455.1 | - | 4 | 423 | |||
AK313834.1 | - | 5 | 511 | |||
BC009898.2 | - | 5 | 502 | 3506022 | ||
BC085621.1 | - | 5 | 471 | 5540463 | ||
Z49099.1 | - | 5 | 455 |