Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006284.3 | NP_006275.1 | 5 | 688 | ||
GenBank mRNA | AL833496.1 | - | 3 | 743 | ||
BC012088.1 | - | 5 | 645 | 4562773 | ||
BM990572.1 | - | 1 | 143 | |||
CR541943.1 | - | 5 | 633 | |||
U13991.1 | - | 5 | 650 |