Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003195.4 | NP_003186.1 | 3 | 803 | ||
NM_198723.1 | NP_942016.1 | 4 | 715 | |||
XM_005260229.3 | XP_005260286.1 | 3 | 945 | |||
XM_011529022.1 | XP_011527324.1 | 7 | 2431 | |||
XM_011529023.1 | XP_011527325.1 | 7 | 2373 | |||
XM_011529027.2 | XP_011527329.1 | 5 | 875 | |||
XM_017028036.1 | XP_016883525.1 | 7 | 2431 | |||
XM_017028037.1 | XP_016883526.1 | 5 | 875 | |||
GenBank mRNA | AJ844626.2 | - | 1 | 23 | ||
AK027824.1 | - | 4 | 508 | |||
AK226140.1 | - | 1 | 1054 | |||
BC018896.2 | - | 3 | 249 | 3955723 | ||
BC050623.1 | - | 3 | 302 | 5171174 | ||
BI668232.1 | - | 3 | 321 | |||
BI756937.1 | - | 4 | 486 | |||
CR456940.1 | - | 3 | 147 | |||
D50495.1 | - | 3 | 165 |