Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003218.3 | NP_003209.2 | 9 | 2143 | ||
NM_017489.2 | NP_059523.2 | 10 | 2203 | |||
XM_005251291.3 | XP_005251348.1 | 11 | 2291 | |||
XM_005251292.3 | XP_005251349.1 | 10 | 2231 | |||
XM_011517582.2 | XP_011515884.1 | 12 | 2358 | |||
XM_017013792.1 | XP_016869281.1 | 10 | 2041 | |||
GenBank mRNA | AF003001.1 | - | 9 | 2131 | ||
BF056993.1 | - | 1 | 500 | |||
U40705.1 | - | 10 | 2191 |