Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRO SIRNA Assays ›
Gene Symbol:
TRO
Gene Name:
trophinin
Gene Aliases:
MAGE-d3, MAGED3
Chromosome Location:
Chr.X: 54920563 - 54931436 on Build GRCh38
UniGene ID:
Hs.633653
Species:
Human
siRNA ID
HSS144343
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRO
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039705.2
NP_001034794.1
4
1623
NM_001271183.1
NP_001258112.1
2
343
NM_001271184.1
NP_001258113.1
3
243
NM_016157.3
NP_057241.2
4
1623
NM_177556.2
NP_808224.1
4
1608
NM_177557.2
NP_808225.1
3
417
NR_073148.1
4
1608
NR_073149.1
3
417
XM_006724600.2
XP_006724663.1
4
1389
XM_011530808.1
XP_011529110.1
4
1434
XM_011530809.1
XP_011529111.1
4
1385
XM_011530811.2
XP_011529113.1
4
1771
XM_011530812.1
XP_011529114.1
4
1372
XM_011530813.1
XP_011529115.1
4
1422
XM_011530814.1
XP_011529116.1
4
1316
XM_017029767.1
XP_016885256.1
4
1786
XM_017029768.1
XP_016885257.1
4
1403
XM_017029769.1
XP_016885258.1
4
1385
XM_017029770.1
XP_016885259.1
4
1434
XM_017029771.1
XP_016885260.1
4
1762
XM_017029772.1
XP_016885261.1
4
1403
XM_017029773.1
XP_016885262.1
4
1314
GenBank mRNA
AB029037.2
-
4
1344
AF349719.1
-
4
1355
AF349720.1
-
4
1355
AF349721.1
-
3
164
AF349722.1
-
3
164
AK001300.1
-
2
1053
AK027489.1
-
4
1362
AK225841.1
-
3
216
AK296329.1
-
2
82
AK299568.1
-
3099
BC026914.1
-
4
1338
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
embryo implantation
negative regulation of cell growth
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline